missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF800 (aa 535-654) Control Fragment Recombinant Protein

Catalog No. RP104939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104939 Supplier Invitrogen™ Supplier No. RP104939
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65708 (PA5-65708. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May be involved in transcriptional regulation.

Specifications

Accession Number Q2TB10
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 168850
Name Human ZNF800 (aa 535-654) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias PP902; Zinc finger protein 800; ZNF800
Common Name ZNF800
Gene Symbol ZNF800
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IRHITVVHKKSSRYLGKITASLEIRAIKKPIDFVLNKVAKRGPSRDEAKHSDSKHDGTSNSPSKKYEVADVGIEVKVTKNFSLHRCNKCGKAFAKKTYLEHHKKTHKANASNSPEGNKTK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less