missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human VEGF R2/KDR Quantikine ELISA Kit, R&D Systems

$676.55 - $3,247.17

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Gene ID (Entrez) 3791
Sample Type Cell Culture Supernates, Cell Lysates, Serum, EDTA Plasma, Heparin Plasma
Sample Volume Cell Culture Supernates (100μL), Cell Lysates (100μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL)
Species Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
DVR200 R & D Systems
RDSDVR200
1 Plate Each for $676.55
SVR200 R & D Systems
RDSSVR200
6 Plates Each for $3,247.17
Specifications

Description

VEGF R1 (Flt-1), VEGF R2 (KDR/Flk-1), and VEGF R3 (Flt-4) belong to the class III subfamily of receptor tyrosine kinases (RTKs). All three receptors contain seven immunoglobulin-like repeats in their extracellular domain and kinase insert domains in their intracellular region. They are best known for regulating VEGF family-mediated vasculogenesis, angiogenesis, and lymphangiogenesis. They are also mediators of neurotrophic activity and regulators of hematopoietic development. VEGF R2 is thought to be the primary inducer of VEGF-mediated blood vessel growth, while VEGF R3 plays a significant role in VEGF-C and VEGF-D-mediated lymphangiogenesis.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Cell Lysates (100μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL)
Sensitivity: 11.4pg/mL
Specificity: Natural and recombinant human VEGF R2
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Cell Culture Supernates, Cell Lysates, Serum, EDTA Plasma, Heparin Plasma
Human
3791
Cell Culture Supernates (100μL), Cell Lysates (100μL), Serum (20μL), EDTA Plasma (20μL), Heparin Plasma (20μL)