Please login to your online account to display your discounted pricing

eBiosciencetrade; Human TSP-1 Platinum ELISA Kit

For the Detection of human Thrombospondin-1 (TSP-1)

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Thrombospondin 1, THBS1, THBS, THBS-1, TSP, TSP-1, TSP1
Target TSP-1
Sample Type Serum and plasma (citrate, heparin, EDTA)
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TSP-1
Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125334

 
ebioscience
LSBMS2100
96 tests Each for $651.05

501125335

 
ebioscience
LSBMS2100TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Thrombospondin 1, THBS1, THBS, THBS-1, TSP, TSP-1, TSP1
Target TSP-1
Sample Type Serum and plasma (citrate, heparin, EDTA)
Sensitivity 0.044ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.