Please login to your online account to display your discounted pricing

Human TSLP DuoSet, R&D Systems

$828.14 - $828.14

Specifications

Applications Development of sandwich ELISA to measure natural and recombinant human TSLP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 kit
Species Human
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

DY1398

 
r & d systems
RDSDY1398
15 96-well plates Each for $828.14
Description & Specifications

Specifications

Applications Development of sandwich ELISA to measure natural and recombinant human TSLP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA kit
Quantity 1 kit
Species Human

Assay Type: ELISA Development Kit
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.