Please login to your online account to display your discounted pricing

Human Total MMP-8 Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSDMP800

 View more versions of this product

Catalog No. DMP800

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4317
Packaging Quantity 1 Plate
Quantity 1 Kit
Sensitivity 0.06ng/mL
Sample Type Cell Culture Supernates, Serum, Heparin Plasma, Saliva
Sample Volume Cell Culture Supernates (10μL), Serum (10μL), Heparin Plasma (10μL), Saliva (10μL)
Species Human
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate