missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human Total MMP-8 Quantikine ELISA Kit, R&D Systems

$624.40 - $2,985.15

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Cell Culture Supernates (10μL), Serum (10μL), Heparin Plasma (10μL), Saliva (10μL)
Species Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
DMP800 R & D Systems
RDSDMP800
1 Plate Each for $624.40
SMP800 R & D Systems
RDSSMP800
6 Plates Each for $2,985.15
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (10μL), Serum (10μL), Heparin Plasma (10μL), Saliva (10μL)
Sensitivity: 0.06ng/mL
Specificity: Natural and recombinant human MMP-8
Cross Reactivity: Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Solid Phase Sandwich ELISA
Cell Culture Supernates (10μL), Serum (10μL), Heparin Plasma (10μL), Saliva (10μL)
Human