Please login to your online account to display your discounted pricing

Human Total M-CSF R DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC3292

 View more versions of this product

Catalog No. DYC3292

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Gene ID (Entrez) 1436
Product Type ELISA Development Kit for intracellular proteins
Packaging Quantity 2 Plates
Quantity 1 Kit
Species Human