Please login to your online account to display your discounted pricing

Human Total Leptin R DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC3892

 View more versions of this product

Catalog No. DYC3892

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3953
Product Type ELISA Development Kit for intracellular proteins
Packaging Quantity 2 Plates
Quantity 1 Kit
Species Human
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.