Please login to your online account to display your discounted pricing

Human Total HO-1/HMOX1 DuoSet IC, R&D Systems

$419.81 - $1,780.05

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 3162
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC3776E

 
r & d systems
RDSDYC3776E
15 Plates Each for $1,780.05

DYC37762

 
r & d systems
RDSDYC37762
2 Plates Each for $419.81

DYC37765

 
r & d systems
RDSDYC37765
5 Plates Each for $828.14
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 3162
Species Human

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.