Please login to your online account to display your discounted pricing

Human Total ErbB3/Her3 DuoSet IC, R&D Systems

$428.87 - $846.01

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 2065
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DYC2342

 
r & d systems
RDSDYC2342
2 Plates Each for $428.87

DYC2345

 
r & d systems
RDSDYC2345
5 Plates Each for $846.01
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Development Kit for intracellular proteins
Quantity 1 Kit
Gene ID (Entrez) 2065
Species Human

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.