Please login to your online account to display your discounted pricing

Human Total Dtk DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC56002

 View more versions of this product

Catalog No. DYC56002

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7301
Packaging Quantity 2 Plates
Product Type ELISA Development Kit for intracellular proteins
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Quantity 1 Kit
Species Human