Please login to your online account to display your discounted pricing

Human Total CD117/c-kit DuoSet IC, R&D Systems


Manufacturer: r & d systems  RDSDYC3322

 View more versions of this product

Catalog No. DYC3322

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3815
Product Type ELISA Development Kit for intracellular proteins
Packaging Quantity 2 Plates
Quantity 1 Kit
Species Human

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.