Please login to your online account to display your discounted pricing

eBioscience™ Human TNF-beta Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human TNF-beta.

$665.55 - $4,197.75

Specifications

For Use With (Application) ELISA
Sample Size 100μL
Synonym Lymphotoxin-alpha, TNF superfamily member 1
Target TNF beta
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TNF-beta
HRP-Conjugate anti-human TNF-beta monoclonal antibody
Human TNF-beta Standard concentrate, 1ng/mL upon dilution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125255

 
life technologies
LSBMS202CE
96 tests Each for $665.55

501125258

 
life technologies
LSBMS202TENCE
10 x 96 tests Each for $4,197.75
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 100μL
Synonym Lymphotoxin-alpha, TNF superfamily member 1
Target TNF beta
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma
Sensitivity 4.6pg/mL
Species Human

The assay detects recombinant, unglycosylated lymphotoxin with the same sensitivity as the natural, glycosylated protein, shows good correlation with the standard cytotoxicity bioassay, and is specific for biologically active TNF-beta without TNF-alpha cross reactivity.