Please login to your online account to display your discounted pricing

Human TNF-alpha Quantikine HS ELISA Kit, R&D Systems

Manufacturer: r & d systems  RDSHSTA00D

 View all options for this product

Catalog No. HSTA00D


Description & Specifications


Gene ID (Entrez) 7124
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 1 Plate
Quantity 1 Kit
Sensitivity 0.191pg/mL
Sample Type Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Sample Volume Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Species Human
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 0.191pg/mL
Specificity: Natural and recombinant human TNF-alpha
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.