Please login to your online account to display your discounted pricing

Human TNF-alpha Quantikine HS ELISA Kit, R&D Systems

$780.83 - $3,747.74

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

HSTA00D

 
r & d systems
RDSHSTA00D
1 Plate Each for $780.83

SSTA00D

 
r & d systems
RDSSSTA00D
6 Plates Each for $3,747.74
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Species Human

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 0.191pg/mL
Specificity: Natural and recombinant human TNF-alpha
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.