Please login to your online account to display your discounted pricing

Human TNF-alpha Quantikine HS ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSSTA00D

 View more versions of this product

Catalog No. SSTA00D

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7124
Product Type Solid Phase Sandwich ELISA
Packaging Quantity 6 Plates
Quantity 1 Pk.
Sensitivity 0.191pg/mL
Sample Type Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Sample Volume Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Species Human
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 0.191pg/mL
Specificity: Natural and recombinant human TNF-alpha
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.