Please login to your online account to display your discounted pricing

Human TNF-alpha Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSDTA00C

 View more versions of this product

Catalog No. DTA00C

  • / Each
 

Description & Specifications

Specifications

Packaging Quantity 1 Plate
Specificity Natural and recombinant human TNF-alpha
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Product Type Solid Phase Sandwich ELISA
Species Human
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Sample Volume Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity 5.5pg/mL
Gene ID (Entrez) 7124
Quantity 1 Kit