missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human TNF-alpha Quantikine ELISA Kit, R&D Systems

$624.40 - $2,985.15

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 7124
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Sample Volume Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
DTA00C R & D Systems
RDSDTA00C
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $624.40
STA00C R & D Systems
RDSSTA00C
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $2,985.15
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Sensitivity: 5.5pg/mL
Specificity: Natural and recombinant human TNF-alpha
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
7124
Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Human
Solid Phase Sandwich ELISA
Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
5.5pg/mL
Natural and recombinant human TNF-alpha