Please login to your online account to display your discounted pricing

Human TNF-alpha Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSTA00C

 View more versions of this product

Catalog No. STA00C

  • / Each
 

Description & Specifications

Specifications

Packaging Quantity 6 Plates
Sample Volume Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL), Citrate Plasma (200μL)
Gene ID (Entrez) 7124
Quantity 1 Pk.
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Specificity Natural and recombinant human TNF-alpha
Sensitivity 5.5pg/mL
Species Human
Product Type Solid Phase Sandwich ELISA
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates