Please login to your online account to display your discounted pricing

eBiosciencetrade; Human TNF-alpha Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human TNF-alpha.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Cachectin, Tumor necrosis factor alpha, DIF
Target TNF alpha
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TNF-alpha
Biotin-Conjugate anti-human TNF-alpha polyclonal antibody
Streptavidin-HRP
Human TNF-alpha Standard lyophilized, 1000pg/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125377

 
ebioscience
LSBMS2234CE
96 tests Each for $665.55

501125378

 
ebioscience
LSBMS2234TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym Cachectin, Tumor necrosis factor alpha, DIF
Target TNF alpha
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 2.3pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.