missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Tumor Necrosis Factor Alpha Recombinant, Thermo Scientific™

Tumor Necrosis Factor Alpha Recombinant protein STNFA from Thermo Scientific™ specifically detects Tumor Necrosis Factor Alpha in samples, and it is validated for ELISA and WB

$170.10 - $2,436.00

Specifications

Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Dependent upon sample type and level (see instructions)
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Target Natural and recombinant human TNFa (human tumor necrosis factor alpha)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Sufficient For Price Quantity  
Catalog Number Mfr. No. Description Sufficient For Price Quantity  
ENEH3TNFA Thermo Scientific™
PIEH3TNFA
Human TNF alpha ELISA Kit 96 microplate wells Each for $651.05
ENEH3TNFA2 Thermo Scientific™
EH3TNFA2
Human TNF alpha ELISA Kit 2 x 96 microplate wells Each for $1,724.10
ENEH3TNFA5 Thermo Scientific™
PIEH3TNFA5
Human TNF alpha ELISA Kit 5 x 96 microplate wells Each for $2,436.00
ENSTNFA Thermo Scientific™
STNFA
Human TNF alpha Standard for ELISA ELISA standard curve with replicates Each for $170.10
Specifications

Description

Tumor Necrosis Factor Alpha Recombinant protein STNFA from Thermo Scientific™ specifically detects Tumor Necrosis Factor Alpha in samples, and it is validated for ELISA and WB

DSCD75 is a 208 amino acid membrane protein with one transmembrane helix. Current testing suggests that the cDNA of DSCD75 may be expressed by osteogenic human mesenchymal stem cells. DSCD75 may be involved in some transporter activities.
Host Species: E. Coli
Formulation: 1 Vial purified Protein (2.5 ng/ml) in 4% BSA and no preservative;

PA5-32685 detects C8orf55 in Human samples. PA5-32685 has been successfully used in Immunohistochemistry and Immunohistochemistry (Paraffin) procedures. PA5-32685 The immunogen is Synthetic 18 amino acid peptide from internal region of human C8orf55..
Description & Specifications

Specifications

Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Dependent upon sample type and level (see instructions)
Natural and recombinant human TNFa (human tumor necrosis factor alpha)
Absorbance at 450nm minus absorbance at 550nm
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)