Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Tumor Necrosis Factor Alpha Recombinant, Thermo Scientific™


Tumor Necrosis Factor Alpha Recombinant protein STNFA from Thermo Scientific™ specifically detects Tumor Necrosis Factor Alpha in samples, and it is validated for ELISA and WB

Manufacturer: thermo scientific™  STNFA

 View more versions of this product

Catalog No. ENSTNFA

  • / Each
 

Description & Specifications

Specifications

Sensitivity <2 pg/mL
Formulation 1 Vial purified Protein (2.5 ng/ml) in 4% BSA and no preservative;
Quantity 1Vial
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Assay Range 15.6 to 1000 pg/mL
Precision Dependent upon sample type and level (see instructions)
Measurement Absorbance at 450nm minus absorbance at 550nm
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Sample Size 50 µL
Specificity STNFA
Sufficient For ELISA standard curve with replicates
Total Assay Time 3.5 hrs
Target Natural and recombinant human TNFa (human tumor necrosis factor alpha)
Description Human TNF alpha Standard for ELISA

Tumor Necrosis Factor Alpha Recombinant protein STNFA from Thermo Scientific™ specifically detects Tumor Necrosis Factor Alpha in samples, and it is validated for ELISA and WB

DSCD75 is a 208 amino acid membrane protein with one transmembrane helix. Current testing suggests that the cDNA of DSCD75 may be expressed by osteogenic human mesenchymal stem cells. DSCD75 may be involved in some transporter activities.
Host Species: E. Coli
Formulation: 1 Vial purified Protein (2.5 ng/ml) in 4% BSA and no preservative;

PA5-32685 detects C8orf55 in Human samples. PA5-32685 has been successfully used in Immunohistochemistry and Immunohistochemistry (Paraffin) procedures. PA5-32685 The immunogen is Synthetic 18 amino acid peptide from internal region of human C8orf55..