Please login to your online account to display your discounted pricing

eBiosciencetrade; Human TIE-2 Platinum ELISA Kit

For the quantitative detection of human TIE-2.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym CD202b antigen, VMCM1, TEK
Target CD202b (TIE2)
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human TIE-2
Biotin-Conjugate anti-human TIE-2 polyclonal antibody
Streptavidin-HRP
Human TIE-2 Standard lyophilized, 20ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5017950

 
ebioscience&trade
LSBMS2042
96 tests Each for $651.05

5017951

 
ebioscience&trade
LSBMS2042TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym CD202b antigen, VMCM1, TEK
Target CD202b (TIE2)
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.03ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.