missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Human TGF-beta1 Platinum ELISA Kit

For the quantitative detection of human TGF-beta1.

Manufacturer:  Invitrogen™ LSBMS2494TEN

 View more versions of this product

Catalog No. 501125413



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TGF-beta1
Biotin-Conjugate anti-human TGF-beta1 monoclonal antibody
Streptavidin-HRP
Human TGF-beta1 Standard lyophilized, 4ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

ELISA
8.6pg/mL
40μL (pretreated)
TGF beta 1
10 x 96 tests
Cell culture supernatant (w/o FCS), serum, plasma (EDTA, citrate, heparin)
Human
Camurati-Engelmann disease, DPD1
Documents