Please login to your online account to display your discounted pricing

eBioscience™ Human TGF beta 2, Recombinant Protein, Carrier-Free

Human Recombinant Protein

$1,898.05 - $1,898.05

Specifications

Name TGF beta 2
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123507

 
life technologies
LS34847882
100μg Each for $1,898.05
Description & Specifications

Specifications

Name TGF beta 2
Recombinant Yes
Conjugate Unlabeled
Purity >98%
Purification Method SDS-PAGE
Source E. coli
For Use With (Application) Functional Assays
Molecular Weight 25kDa
Storage Requirements Store at less than or equal to -70°C.

Human Transforming Growth Factor beta 2 (TGF beta 2) is a member of a superfamily of disulfide-linked homodimeric proteins secreted as latent proteins and stored at the cell surface and extracellular matrix. The bioactive TGF beta 2 is released from a latent complex by proteolytic processing and conformation changes. TGF beta 2 regulates cell proliferation, growth, differentiation and motility as well as synthesis and deposition of the extracellular matrix and is also involved in embryogenesis, tissue remodeling and wound healing.