Please login to your online account to display your discounted pricing

Human TGF-beta 2 Quantikine ELISA Kit, R&D Systems

$662.25 - $3,178.57

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Cell Culture Supernates (125μL), Serum (125μL), EDTA Plasma (125μL), Heparin Plasma (125μL), Citrate Plasma (125μL)
Species Human
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DB250

 
r & d systems
RDSDB250
1 Plate Each for $662.25

SB250

 
r & d systems
RDSSB250
6 Plates Each for $3,178.57
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Volume Cell Culture Supernates (125μL), Serum (125μL), EDTA Plasma (125μL), Heparin Plasma (125μL), Citrate Plasma (125μL)
Species Human

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (125μL), Serum (125μL), EDTA Plasma (125μL), Heparin Plasma (125μL), Citrate Plasma (125μL)
Sensitivity: 7pg/mL
Specificity: Natural and recombinant human TGF-beta 2 and natural porcine TGF-beta 2.
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.