Please login to your online account to display your discounted pricing

Human TGF-beta 1 Quantikine ELISA Kit, R&D Systems


A 4.5 hour solid phase ELISA designed to measure TGF-beta 1 in acid activated cell culture supernates, serum, plasma, and urine. It contains recombinant human TGF-beta 1 expressed by CHO cells

Manufacturer: r & d systems  RDSDB100B

 View more versions of this product

Catalog No. DB100B

  • / Each
 

Description & Specifications

Specifications

Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Urine
Product Type Solid Phase Sandwich ELISA
Sensitivity 15.4pg/mL
Sample Volume Cell Culture Supernates (100μL), Serum (40μL), EDTA Plasma (40μL), Urine (100μL)
Packaging Quantity 1 Plate
Quantity 1 Kit
Cross Reactivity Cross-species reactivity observed with 1 or more species tested. Cross-reactivity observed with 1 or more available related molecules
Species Human
Gene ID (Entrez) 7040
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Serum (40μL), EDTA Plasma (40μL), Urine (100μL)
Sensitivity: 15.4pg/mL
Specificity: Natural and recombinant TGF-beta 1. This kit also recognizes human TGF-beta 1.2 and mouse, rat, porcine, and canine TGF-beta 1.
Cross Reactivity: Cross-species reactivity observed with 1 or more species tested. Cross-reactivity observed with 1 or more available related molecules.