Please login to your online account to display your discounted pricing

Human sVEGF R1/Flt-1 Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSDVR100B

 View more versions of this product

Catalog No. DVR100B

  • / Each
 

Description & Specifications

Specifications

Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Saliva, Follicular Fluid
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Packaging Quantity 1 Plate
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Saliva (100μL), Follicular Fluid (100μL)
Gene ID (Entrez) 2321
Species Human
Quantity 1 Kit