Please login to your online account to display your discounted pricing

Human sVEGF R1/Flt-1 Quantikine ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSVR100B

 View more versions of this product

Catalog No. SVR100B

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2321
Packaging Quantity 6 Plates
Quantity 1 Pk.
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Saliva, Follicular Fluid
Sample Volume Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Saliva (100μL), Follicular Fluid (100μL)
Species Human
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.