Please login to your online account to display your discounted pricing

eBiosciencetrade; Human sVCAM-1 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sVCAM-1.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 100μL (1:50 prediluted)
Synonym Vascular cell adhesion protein 1, CD106 antigen
Target CD106 (VCAM-1), sCD106 (sVCAM-1)
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sVCAM-1
Conjugate Mixture containing Biotin-Conjugate anti-human sVCAM-1 monoclonal antibody mixed with Streptavidin-HRP
Human sVCAM-1 Standard lyophilized, 200ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125393

 
ebioscience
LSBMS232CE
96 tests Each for $665.55

501125394

 
ebioscience
LSBMS232TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 100μL (1:50 prediluted)
Synonym Vascular cell adhesion protein 1, CD106 antigen
Target CD106 (VCAM-1), sCD106 (sVCAM-1)
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.6ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.