Please login to your online account to display your discounted pricing

Human sVCAM-1/CD106 Quantikine ELISA Kit, R&D Systems

$676.55 - $3,247.17

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 7412
Sample Volume Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Sensitivity 1.26ng/mL
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity    

DVC00

 
r & d systems
RDSDVC00
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $676.55

SVC00

 
r & d systems
RDSSVC00
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $3,247.17
Description & Specifications

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 7412
Sample Volume Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Sensitivity 1.26ng/mL
Species Human

Human VCAM-1 (CD106), a member of the immunoglobulin superfamily, is a cell surface protein expressed by activated endothelial cells and certain leukocytes such as macrophages. VCAM-1 expression is induced by IL-1beta, IL-4, TNF-alpha and IFN-gamma. VCAM-1 binds to leukocyte integrins VLA-4 and Integrin alpha 4 beta 7.

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL)
Sensitivity: 1.26ng/mL
Specificity: Natural and recombinant human sVCAM-1
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.