Please login to your online account to display your discounted pricing

eBioscience™ Human sVAP-1 Platinum ELISA Kit

For the quantitative detection of human sVAP-1.

$520.84 - $2,932.34

Specifications

For Use With (Application) ELISA
Sample Size 10μL (1:100 prediluted)
Synonym Vascular adhesion protein 1, copper-containing 3
Target sVAP-1
Conjugate Biotin
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sVAP-1
Biotin-Conjugate anti-human sVAP-1 monoclonal antibody
Streptavidin-HRP
Human sVAP-1 Standard lyophilized, 4ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018183

 
life technologies
LSBMS259
96 tests Each for $520.84

5018184

 
life technologies
LSBMS259TEN
10 x 96 tests Each for $2,932.34
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL (1:100 prediluted)
Synonym Vascular adhesion protein 1, copper-containing 3
Target sVAP-1
Conjugate Biotin
Sample Type Cell culture supernatant, serum, plasma (EDTA, heparin), amniotic fluid
Sensitivity 19pg/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.