Please login to your online account to display your discounted pricing

eBiosciencetrade; Human sTNF-R (80 kd) Platinum ELISA Kit, for in vitro diagnostic use

Provides a simple, rapid and highly sensitive method for the determination of soluble TNF-R (80 kd) levels in body fluids or cell culture supernatants.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Tumor necrosis factor receptor 2, TNFR2, CD120b
Target sTNF Receptor, TNF Receptor
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sTNF-R (80 kDa)
HRP-Conjugate anti-human sTNF-R (80 kDa) monoclonal antibody
Human sTNF-R (80 kDa) Standard, 10ng/mL
Control, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125340

 
ebioscience
LSBMS211CE
96 tests Each for $665.55

501125341

 
ebioscience
LSBMS211TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Tumor necrosis factor receptor 2, TNFR2, CD120b
Target sTNF Receptor, TNF Receptor
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), urine
Sensitivity 0.10ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.