Please login to your online account to display your discounted pricing

eBioscience™ Human sTNF-R (60 kd) Platinum ELISA Kit

For the quantitative detection of human sTNF-R (60 kd).

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Tumor necrosis factor receptor 1, TNF-R1, CD120a
Target sTNF Receptor, TNF Receptor
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal polyclonal antibody to human sTNF-R (60 kDa)
HRP-Conjugate anti-human sTNF-R (60 kDa) monoclonal antibody
Human sTNF-R (60 kDa) Standard, 10ng/mL upon dilution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5017918

 
life technologies
LSBMS203
96 tests Each for $651.05

5017937

 
life technologies
LSBMS203TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Tumor necrosis factor receptor 1, TNF-R1, CD120a
Target sTNF Receptor, TNF Receptor
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.05ng/mL
Species Human

  • Provides a simple, rapid and highly sensitive method for the determination of soluble TNF-R (60 kd) levels in body fluids or cell culture supernatants
  • This assay will help to clarify the possible diagnostic and prognostic value of circulating sTNF-R (60 kd) in various neoplastic and inflammatory diseases