missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SRP54 (aa 169-248) Control Fragment Recombinant Protein

Catalog No. RP105221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105221 Supplier Invitrogen™ Supplier No. RP105221
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66449 (PA5-66449. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).

Specifications

Accession Number P61011
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6729
Name Human SRP54 (aa 169-248) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 54 K subunit of signal recognition particle; 54 kDa; signal recognition particle 54; signal recognition particle 54 kDa protein; signal recognition particle 54 A; signal recognition particle 54 kD; signal recognition particle 54 kDa; SRP54; Srp54a
Common Name SRP54
Gene Symbol SRP54
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VIIASEGVEKFKNENFEIIIVDTSGRHKQEDSLFEEMLQVANAIQPDNIVYVMDASIGQACEAQAKAFKDKVDVASVIVT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less