Please login to your online account to display your discounted pricing

eBioscience Human sP-selectin Platinum ELISA Kit

For the quantitative detection of human sP-selectin. P-selectin belongs to the selectin family of adhesion molecules.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Granule membrane protein 140, PADGEM, CD62P
Target CD62P (P-Selectin), sCD62P (sP-Selectin)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sP-selectin
HRP-Conjugate anti-human sP-selectin monoclonal antibody
Human sP-selectin Standard lyophilized, 80ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018042

 
ebioscience
LSBMS2194TEN
10 x 96 tests Each for $4,189.05

5018040

 
ebioscience
LSBMS2194
96 tests Each for $651.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Granule membrane protein 140, PADGEM, CD62P
Target CD62P (P-Selectin), sCD62P (sP-Selectin)
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.20ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.