missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC7A8 (aa 1-40) Control Fragment Recombinant Protein

Catalog No. RP104033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104033 Supplier Invitrogen™ Supplier No. RP104033
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62519 (PA5-62519. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SLC7A8 is sodium-independent, high-affinity transport of large neutral amino acids. It has higher affinity for L-phenylalanine than LAT1 but lower affinity for glutamine and serine. L-alanine is transported at physiological concentrations. SLC7A8 also plays a role in basolateral (re)absorption of neutral amino acids.

Specifications

Accession Number Q9UHI5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23428
Name Human SLC7A8 (aa 1-40) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4F2-LC5; 4F2LC-5; AA408822; cationic amino acid transporter y+ system; glycoprotein-associated amino acid transporter LAT2; hLAT2; integral membrane protein E16H; Large neutral amino acids transporter small subunit 2; LAT2; Lat4; LPI-PC1; L-type amino acid transporter 2; mLAT2; Slc7a8; solute carrier family 7 (amino acid transporter light chain, L system), member 8; solute carrier family 7 (amino acid transporter, L-type), member 8; solute carrier family 7 (cationic amino acid transporter, y+ system), member 8; solute carrier family 7 member 8; solute carrier family 8 (cationic amino acid transporter, y+ system), member 7
Common Name SLC7A8
Gene Symbol SLC7A8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MEEGARHRNNTEKKHPGGGESDASPEAGSGGGGVALKKEI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less