missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human sL-Selectin/CD62L ELISA Kit, R&D Systems

$2,901.06 - $2,901.06

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 6402
Sample Volume Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)
Sensitivity 0.3ng/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
SBBE4B R & D Systems
RDSSBBE4B
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $2,901.06
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)
Sensitivity: 0.3ng/mL
Specificity: Natural and recombinant human sL-Selectin
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
6402
0.3ng/mL
Solid Phase Sandwich ELISA
Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)
Human