Please login to your online account to display your discounted pricing

Human sL-Selectin/CD62L ELISA Kit, R&D Systems


Manufacturer: r & d systems  RDSSBBE4B

 View more versions of this product

Catalog No. SBBE4B

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 6402
Packaging Quantity 6 Plates
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Sufficient For Sufficient materials to run ELISAs on at least six 96-well plates
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma, Citrate Plasma
Sensitivity 0.3ng/mL
Quantity 1 Pk.
Species Human
Sample Volume Cell Culture Supernates (10μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)