Please login to your online account to display your discounted pricing

eBiosciencetrade; Human sIL-2R Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sIL-2R.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym CD25 antigen, TAC, Interleukin-2 receptor alpha
Target IL-2 Receptor, sIL-2 Receptor
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sIL-2R
HRP-Conjugate anti-human sIL-2R monoclonal antibody
Human sIL-2R Standard lyophilized, 40ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125342

 
ebioscience
LSBMS2122CE
96 tests Each for $665.55

501125343

 
ebioscience
LSBMS2122TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 50μL
Synonym CD25 antigen, TAC, Interleukin-2 receptor alpha
Target IL-2 Receptor, sIL-2 Receptor
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.27ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.