Please login to your online account to display your discounted pricing

eBioscience™ Human sICAM-3 Platinum ELISA Kit

For the quantitative detection of human sICAM-3.

$520.84 - $2,932.34

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym CD50 antigen, Intercellular adhesion molecule 3
Target sCD50 (sICAM-3)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sICAM-3
HRP-Conjugate anti-human sICAM-3 monoclonal antibody
Human sICAM-3 Standard, 50ng/mL
Sample Diluent
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018036

 
life technologies
LSBMS218
96 tests Each for $520.84

5018039

 
life technologies
LSBMS218TEN
10 x 96 tests Each for $2,932.34
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym CD50 antigen, Intercellular adhesion molecule 3
Target sCD50 (sICAM-3)
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, heparin), amniotic fluid, urine
Sensitivity 0.38ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.