Please login to your online account to display your discounted pricing

eBioscience™ Human sICAM-1 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sICAM-1.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Intercellular adhesion molecule 1, CD54 antigen
Target CD54 (ICAM-1), sCD54 (sICAM-1)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sICAM-1
HRP-Conjugate anti-human sICAM-1 monoclonal antibody
Human sICAM-1 Standard, 100ng/mL
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125251

 
life technologies
LSBMS201CE
96 tests Each for $665.55

501125253

 
life technologies
LSBMS201TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 10μL
Synonym Intercellular adhesion molecule 1, CD54 antigen
Target CD54 (ICAM-1), sCD54 (sICAM-1)
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, heparin, citrate)
Sensitivity 2.2ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.