Please login to your online account to display your discounted pricing

eBiosciencetrade; Human sHER-2 Platinum ELISA Kit

For the quantitative detection of human soluble HER-2 protein.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 100μL (1:20 prediluted)
Synonym ERBB2, Herstatin, CD340
Target HER-2, sHER-2
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sHER-2
HRP-Conjugate anti-human sHER-2 monoclonal antibody
Human sHER-2 Standard, 10ng/mL
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125281

 
ebioscience
LSBMS207
96 tests Each for $651.05

5017980

 
ebioscience&trade
LSBMS207TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 100μL (1:20 prediluted)
Synonym ERBB2, Herstatin, CD340
Target HER-2, sHER-2
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, heparin), amniotic fluid
Sensitivity 0.06ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.