Please login to your online account to display your discounted pricing

eBioscience™ Human sE-selectin Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human sE-selectin.

$665.55 - $4,334.05

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym ELAM1, endothelial adhesion molecule 1, CD62E
Target CD62E (E-Selectin), sCD62E (sE-Selectin)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sE-selectin
HRP-Conjugate anti-human sE-selectin monoclonal antibody
Human sE-selectin Standard lyophilized, 100ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125273

 
life technologies
LSBMS205CE
96 tests Each for $665.55

501125275

 
life technologies
LSBMS205TENCE
10 x 96 tests Each for $4,334.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym ELAM1, endothelial adhesion molecule 1, CD62E
Target CD62E (E-Selectin), sCD62E (sE-Selectin)
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.3ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.