missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Human sE-Selectin/CD62E Quantikine ELISA Kit, R&D Systems

a 4.5 hour solid phase ELISA designed to measure sE-Selectin in cell culture supernates, serum, and plasma.

$663.51 - $3,180.69

Specifications

Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Gene ID (Entrez) 6401
Sample Volume Cell Culture Supernates (50μL), Serum (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)
Sensitivity 0.027ng/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Sufficient For Price Quantity  
DSLE00 R & D Systems
RDSDSLE00
1 Plate Sufficient materials to run ELISAs on at least one 96-well plate Each for $663.51
SSLE00 R & D Systems
RDSSSLE00
6 Plates Sufficient materials to run ELISAs on at least six 96-well plates Each for $3,180.69
Specifications

Description

Assay Type: Solid Phase Sandwich ELISA
Sample Type/Volume Required: Cell Culture Supernates (50μL), Serum (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)
Sensitivity: 0.027ng/mL
Specificity: Natural and recombinant human sE-Selectin
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Description & Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
6401
0.027ng/mL
Solid Phase Sandwich ELISA
Cell Culture Supernates (50μL), Serum (10μL), Heparin Plasma (10μL), Citrate Plasma (10μL)
Human