Please login to your online account to display your discounted pricing

eBioscience™ Human sCD93 ELISA Ready-SET-Go!™ Kit

For quantitative measurement of human sCD93

$332.05 - $849.19

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym soluble C1q receptor
Target CD93
Sample Type Cell culture supernatant, serum and plasma
View More Specs
Includes:  Capture Antibody Concentrate
Detection Antibody Concentrate
Human sCD93 Standard (lyophilized): 800ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent Z
Detection enzyme
Tetramethylbenzidine (TMB) Substrate Solution

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    

501125111

 
life technologies
LS885034022
96-well plates 2 x 96 tests Each for $332.05

501125112

 
life technologies
LS885034076
96-well plates 20 x 96 tests Each for $849.19

501125113

 
life technologies
LS885034077
Each for $799.89

501125114

 
life technologies
LS885034086
96-well plates 10 x 96 tests Each for $590.37

501125115

 
life technologies
LS885034088
Each for $565.72
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL
Synonym soluble C1q receptor
Target CD93
Sample Type Cell culture supernatant, serum and plasma
Sensitivity 3.3ng/mL
Species Human

Human sCD93 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA).