missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Human sCD93 ELISA Ready-SET-Go!™ Kit

For quantitative measurement of human sCD93

Manufacturer:  Invitrogen™ LS885034086

 View more versions of this product

Catalog No. 501125114



Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
Human sCD93 Standard (lyophilized): 800ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent Z
Detection enzyme
Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

Human sCD93 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA).
Specifications

Specifications

ELISA
10 x 96 tests
Cell culture supernatant, serum and plasma
Human
soluble C1q receptor
96-well plates
3.3ng/mL
20μL
CD93
Documents

For Research Use Only