Please login to your online account to display your discounted pricing

eBioscience™ Human sCD44var (v6) Platinum ELISA Kit

For the quantitative detection of human sCD44var (v6).

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 20μL (1:20 prediluted)
Synonym VFF-7 Phagocytic glycoprotein I, Hermes antigen
Target sCD44var (v6)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44var (v6)
HRP-Conjugate anti-human sCD44var (v6) monoclonal antibody
Human sCD44var (v6) Standard lyophilized, 20ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5017994

 
life technologies
LSBMS210
96 tests Each for $651.05

5017996

 
life technologies
LSBMS210TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL (1:20 prediluted)
Synonym VFF-7 Phagocytic glycoprotein I, Hermes antigen
Target sCD44var (v6)
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Sensitivity 0.05ng/mL
Species Human

The sCD44var(v6) ELISA detects all circulating CD44 isoforms comprising the sCD44var(v6) sequences