Please login to your online account to display your discounted pricing

eBioscience™ Human sCD44var (v5) Platinum ELISA Kit

For the quantitative detection of human sCD44var (v5).

$520.84 - $2,932.34

Specifications

For Use With (Application) ELISA
Sample Size 20μL (1:6 prediluted)
Synonym VFF-8 Phagocytic glycoprotein I, Hermes antigen
Target sCD44var (v5)
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44var (v5)
HRP-Conjugate anti-human sCD44var (v5) monoclonal antibody
Human sCD44var (v5) Standard lyophilized, 20ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5018044

 
life technologies
LSBMS220
96 tests Each for $520.84

5018046

 
life technologies
LSBMS220TEN
10 x 96 tests Each for $2,932.34
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL (1:6 prediluted)
Synonym VFF-8 Phagocytic glycoprotein I, Hermes antigen
Target sCD44var (v5)
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine
Sensitivity 0.14ng/mL
Species Human

The sCD44var(v5) ELISA detects all circulating CD44 isoforms comprising the sCD44var(v5) sequences

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.