Please login to your online account to display your discounted pricing

eBioscience™ Human sCD44std Platinum ELISA Kit

For the quantitative detection of human sCD44std.

$651.05 - $4,189.05

Specifications

For Use With (Application) ELISA
Sample Size 20μL (1:60 prediluted)
Synonym Phagocytic glycoprotein I, Hermes antigen, Epican
Target CD44std, sCD44std
Conjugate HRP
View More Specs
Includes:  Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD44std
HRP-Conjugate anti-human sCD44std monoclonal antibody
Human sCD44std Standard, 8ng/mL
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5017988

 
life technologies
LSBMS2092
96 tests Each for $651.05

5017991

 
life technologies
LSBMS2092TEN
10 x 96 tests Each for $4,189.05
Description & Specifications

Specifications

For Use With (Application) ELISA
Sample Size 20μL (1:60 prediluted)
Synonym Phagocytic glycoprotein I, Hermes antigen, Epican
Target CD44std, sCD44std
Conjugate HRP
Sample Type Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine
Sensitivity 0.02ng/mL
Species Human

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.